Nylon Cord
Nylon Cord
Nylon Cord
Catalog # 470159-550
Supplier:   Wards
Supplier #:  PH0302A
Nylon Cord
Catalog # 470159-550
Supplier:  Wards
Nylon Cord
Nylon Cord
Nylon Cord
Price
38,70 $
Log in pour la disponibilité
Each
Product Details & Documents
A Necessity for the Lab.
  • Suitable for small pulley system and pendulums
Length is approximately 30 meters.
Spécifications
  • Désignation:
    Nylon Cord