Oxoid VCAT Selective Supplement, Hardy Diagnostics
Catalog # CA89423-162
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Description:VCAT Selective Supplement
- Size:10 vials
- Cat. no.:CA89423-162
- Supplier No.:SR104B
Specifications
About this item
An antibiotic selective supplement for culture media, for the isolation of Neisseria gonorrhoeae
An antibiotic selective supplement for culture media, for the isolation of Neisseria gonorrhoeae, such as Thayer Martin. Contains Vancomycin, Colistin, Anisomysin, and Trimethoprim.