Trio.Bas™ Gas System with Aspirating Chamber Kit
Supplier: HARDY DIAGNOSTICS CA
Certificates
About this item
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
- Test is manual and the time is calculated by a timer
- Each component of the Trio.Gas™ adapter instrument is autoclavable
- Calibrated regulator guarantees a 100 liters per minute flow rate
The Trio.Bas™ microbial compressed gas system is designed to test the presence of microorganisms in compressed gas samples supplied from tanks and pipes under pressure.
If automatically timed samples and electronic data records are required, the MINI or MONO instrument (76196-430, 76196-732, 76196-438, 76196-440) used concurrently with the GAS system is necessary.
Delivery information: Trio.Bas™ Gas System Kit Contains:
Trio.Gas™ System, high pressure gas sampler attachment
Includes stainless steel electrovalve and fixing system for gas connection
Aspirating gas chamber; easily aseptically connected to the output of the gas system
Stainless steel aspirating head (choose Contact or Petri plate), autoclavable
Stainless steel cover head; protection cover for aspirating head, autoclavable
Robustus hard shell carrying case to protect Trio.Gas™ system in transit
IQ, OQ, PQ, fillable documents
Trio.Gas™ System, high pressure gas sampler attachment
Includes stainless steel electrovalve and fixing system for gas connection
Aspirating gas chamber; easily aseptically connected to the output of the gas system
Stainless steel aspirating head (choose Contact or Petri plate), autoclavable
Stainless steel cover head; protection cover for aspirating head, autoclavable
Robustus hard shell carrying case to protect Trio.Gas™ system in transit
IQ, OQ, PQ, fillable documents