Saline, 0.85%
Saline, 0.85%
Saline, 0.85%
Supplier:  Hardy Diagnostics
Certificates
Saline, 0.85%
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
For the preparation of isotonic suspensions and dilutions of microorganisms.
Saline products are recommended for procedures that require the use of an isotonic diluent. It is also recommended foruse in preparing test suspensions of organisms.
Product Family Options
Product Information
  • Désignation
    Type de conditionnement
    Volume
    Utilisation
    Packed
    Taille
    Availability
    Price
  • Saline, 0.85%
    Glass tube
    1 ml
    Clinical
    20/pack
    13×100 mm
    Log in for availability
    Log in for availability
    Pack of 20 (1ml)
    Pack of 20 (1ml) - $95.10
    $95.10
    Pack of 20 (1ml)
       $95.10
    Minimum order quantity is 1
    Supplier Number: K248
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Saline, 0.85%
    PC bottle
    1 L
    Clinical
    10/pack
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $421.00
    $421.00
    Pack of 10
       $421.00
    Minimum order quantity is 1
    Supplier Number: U157
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Saline, 0.85%
    Glass tube
    4 ml
    Clinical
    20/pack
    13×100 mm
    Log in for availability
    Log in for availability
    Pack of 20 (4ml)
    Pack of 20 (4ml) - $111.00
    $111.00
    Pack of 20 (4ml)
       $111.00
    Supplier Number: R47
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Saline, 0.85%
    Glass tube
    9 ml
    Clinical
    20/pack
    16×100 mm
    Log in for availability
    Log in for availability
    Each(9ml)
    Each(9ml) - $119.00
    $119.00
    Each(9ml)
       $119.00
    Minimum order quantity is 1
    Supplier Number: K52
  • Saline, 0.85%
    PC bottle
    1 L
    Clinical
    10/pack
    Log in for availability
    Log in for availability
    Each
    Each - $421.00
    $421.00
    Each
       $421.00
    Minimum order quantity is 1
    Supplier Number: U157
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.