Regan-Lowe Agar without Cephalexin, Hardy Diagnostics
Catalog # 75802-938
Supplier: HARDY DIAGNOSTICS CA
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Plate
- Description:Regan-Lowe Agar without Cephalexin
- Size:15 x 100 mm
- Cat. no.:75802-938
- Supplier No.:A63
Specifications
About this item
For the selective isolation of Bordetella pertussis.
(Charcoal Blood Agar with 10% Horse Blood and w/o Cephalexin) For the selective isolation of Bordetella pertussis. Bordatella are respiratory pathogens, they reside on the mucous membranes of the respiratory tract.