Q-Swab™ by Hygiena, Ready-to-Use Sampling Swab with Buffered Peptone Water, Hardy Diagnostics
Catalog # CA10054-176
Supplier: HARDY DIAGNOSTICS CA
Specifications
- Description:Q-Swab™, Ready-to-use Sampling Swab with Buffered Peptone Water
- Size:250 per box
- Cat. no.:CA10054-176
- Supplier No.:QS1000
Specifications
About this item
Q-Swab is a ready-to-use sample collection and delivery device for environmental surface sampling.
- Self-contained, gamma irradiated
- Swab, snap and squeeze, and a sample is ready
- Q-Swab can be used either wet or dry to sample environmental surfaces
Money and time are saved by eliminating pipettes and broth preparation required with other sample collection methods. Each of the broths offered in Q-Swab are self-contained, gamma irradiated and held away from the swab by utilizing patented Snap-Valve technology. When the Snap-Valve is snapped, broth is released down the swab shaft, neutralizing residual sanitizers and facilitating recovery of bacteria. The sample is then ready to be poured onto dehydrated media film or streaked onto agar plates for culturing.