Phenol Red Broth with Dextrose, Hardy Diagnostics
CA10124-528
:
- Description:Phenol red broth with dextrose
- Size:16×125 mm
- Cat. no.:CA10124-528
- Volume:10 ml
- Supplier No.:Y304
For the differentiation of microorganisms by carbohydrate fermentation.
For the differentiation of microorganisms by carbohydrate fermentation. Contains a durham tube for gas detection.