Oxoid Novobiocin Supplement, Hardy Diagnostics
Catalogue # CA89423-158
Fournisseur: HARDY DIAGNOSTICS CA
Numéro CAS:
Spécifications
- Description:Novobiocin Supplement
- Size:10 x 10 mg
- Cat. no.:CA89423-158
- Supplier No.:SR181E
Spécifications
À propos de cet article
A selective supplement for the addition to culture media to enhance inhibition of Gram positive organisms.
A selective supplement for the addition to culture media to enhance inhibition of Gram positive organisms.