Neutralizing Buffered Peptone Water
Used in the recovery of sub-lethally injured Salmonella species from industrial samples prior to selective enrichment and isolation.
This medium is recommended by the USDA FSIS (United States Department of Agriculture, Food Safety and Inspection Service) for poultry verification sampling.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together