LoopCaddy™, Hardy Diagnostics
LoopCaddy, Each
Catalog # CA89423-150
Supplier: HARDY DIAGNOSTICS CA
: K24H
Jump to:
Product Details & Documents
LoopCaddy™, stand that holds disposable loops for ready use.
A convenient, all acrylic loop holder for use with the resealable new Hardy Diagnostics loop pouches.
The resealable loop pouches slide onto the loop holder allowing the user to simply pull out one loop at a time and reseal the package. The free standing loop holder can be placed anywhere and is designed to be very stable. This product makes the laborious task of opening and closing loop containers fast and easy.
Spécifications
- Désignation:LoopCaddy
- Taille:Each