Oxoid Half Fraser Supplement, Hardy Diagnostics
Catalogue # CA89423-176
Fournisseur: HARDY DIAGNOSTICS CA
Numéro CAS:
Spécifications
- Description:Half Fraser Supplement
- Size:10 vials
- Melting point:35-37 °C
- Cat. no.:CA89423-176
- Volume:10× 225 ml
- Supplier No.:SR166E
Spécifications
À propos de cet article
A selective supplement for the isolation of Listeria spp. from food and environmental samples
A selective supplement for the isolation of Listeria spp
Vial contents (each vial is sufficient to supplement 225ml of medium):
Ferric ammonium citrate 112.50mg
Nalidixic acid 2.25mg
Acriflavine hydrochloride 2.8125mg
from food and environmental samples when used with Criterion™ Fraser Broth.