Fluid A, USP
Fluid A, USP
Fluid A, USP
Fluid A, USP
Fluid A, USP
A rinsing and diluting fluid for use in USP testing protocols.
A rinsing and diluting fluid for use in USP testing protocols. Contains peptone 0.1%.
Hardy Diagnostics' sterility media and rinse solutions are manufactured in an ISO 13485 environmentally controlled production center. Each lot is certified for pH, sterility and growth promotion using ATCC® strains specified by the USP. Our approach to manufacturing ensures all sterility media and rinse fluids provide the highest level of clarity for improved reading accuracy, thus significantly reducing the risk of incorrect interpretation and false results.
Ordering information: Order by the pack of 10
Product Family Options
  • Désignation
    Taille
    Volume
    Type de conditionnement
    Minimum Order Quantity
  • Fluid A, USP
    200 ml
    Serum bottle
    10
     : -
  • Fluid A, USP
    200 ml
    Bottle, Boston round
    10
    Pack of 12
    Pack of 12 - $860.00
    $860.00
    Pack of 12
       $860.00
    Minimum order quantity is 10
     : U436
  • Fluid A, USP
    1 L
    1 L
    Bottle
    Pack of 10
    Pack of 10 - $923.00
    $923.00
    Pack of 10
       $923.00
    Minimum order quantity is 1
     : U208