Buffer #6, Hardy Diagnostics
CA10693-108
:
- Pack type:PET bottle
- Description:Buffer No. 6, USP
- Cat. no.:CA10693-108
- Volume:500 ml
- Supplier No.:U556
For use in the determination of antimicrobial activity per USP.
: Minimum order of 4 packs of 10.