HardyDisks™ Bacitracin, Hardy Diagnostics
Catalogue # CA10790-374
Fournisseur: HARDY DIAGNOSTICS CA
Numéro CAS:
Spécifications
- Description:Bacitracin 0.04 unit, HDx
- Size:5 x 50 disks
- Cat. no.:CA10790-374
- Supplier No.:Z7025
Spécifications
À propos de cet article
Used in the presumptive identification of group A, beta-hemolytic streptococci.
Used in the presumptive identification of group A, beta-hemolytic streptococci
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispenser by BD BioSciences.
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
The test differentiates group A streptococci from other Lancefield groups of hemolytic streptococci by inhibiting the growth of group A streptococci around a disk containing approximately 0.04 units bacitracin.