Lambda PFG Ladder, New England Biolabs
Supplier: New England Biolabs (NEB)
Jump to:
About this item
Lambda PFG Ladder consists of one GelSyringe™ dispenser, sufficient for 50 gel lanes
- Suitable for pulsed field gel electrophoresis
- Size range: 48.5 to 1018 kb
- Includes one GelSyringe dispenser
- Suitable for 50 gel lanes
Successively larger concatemers of lambda DNA (cI857 ind 1 Sam7) are embedded in 1% LMP agarose. This product is designed to be used as size markers for pulsed-field gel electrophoresis (PFG).
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Lambda PFG Ladder consists of one GelSyringe™ dispenser, sufficient for 50 gel lanes
- Suitable for pulsed field gel electrophoresis
- Size range: 48.5 to 1018 kb
- Includes one GelSyringe dispenser
- Suitable for 50 gel lanes
Successively larger concatemers of lambda DNA (cI857 ind 1 Sam7) are embedded in 1% LMP agarose. This product is designed to be used as size markers for pulsed-field gel electrophoresis (PFG).
Product Family Options
Product Information
- DésignationTailleAvailabilityPrice
- Supplier Number: N0341SItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: N0341SItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.