Corrugated SwitchBoard™ Anti-Fatigue Matting, Wearwell®
: Wearwell
Corrugated SwitchBoard™ matting prevents life-threatening electrical shocks in high-voltage environments.
- Conforms to ANSI/ASTM D-178-01 Type II, Class 2
- Dielectric strength: 30,000 volts
- Recommended maximum usage per ASTM D178-01: up to 17,000 volts
- Easy to clean corrugated surface design
Designed to protect personnel in areas with dangerous high-voltage equipment. Surface is easily cleaned and will not collect contaminants. Conforms to ANSI/ASTM D-178-01 Type II (a premium quality vinyl compound resistant to UV, ozone and many common chemicals). Ideally suited for use in dry environments.
: Meets ASTM D178-01: 0.224 mm/s. Meets ASTM F1677: Dry COF 0.91.
Frequently Bought Together