Climate Chambers ICH Series, Memmert
Climate Chambers ICH Series, Memmert
   Options durables disponibles
Supplier:   Memmert
Climate Chambers ICH Series, Memmert
Supplier:  Memmert
   Options durables disponibles
Certificates
Climate Chambers ICH Series, Memmert
6 Options Available Below
Product Details & Documents
Memmert stability chambers are constructed of stainless steel inside and out. The ICH L stability chamber will save energy and perform well for any project.
  • From temperatures of at least 10 up to +60 °C including air jacket heating system for gentle all-around heating
  • Humidity range 10 to 80% rh - or rh OFF
  • Illumination unit consisting of 3 fluorescent lights with cold white light (standard illuminant D65, 6.500K) and 2 UV-lamps in the spectral range of 320-400 nm
  • Programmable with internal data logging via included AtmoCONTROL Software
  • Available in 3 sizes - 108 to 749 L volume chambers
Compressor-cooled stability test chambers developed by Memmert stand out due to their unparalleled temperature and humidity homogeneity for long-term stable ambient conditions. The Climate Chamber ICH has been specially designed for testing pharmaceutical products according to ICH, Q1A, and similar global standards for stability tests of cosmetics and food.
These environmentally friendly stability testing chambers are powerful and climate friendly at the same time, they are especially designed for testing pharmaceuticals according o ICH Q1A and Q1B as well as for testing the stability of cosmetics and food. Temperature and humidity are distributed homogeneously and stable throughout the interior.
These incubators have an integrated data logger and digital timer which is adjustable from 1 minute to 99 days and 23 hours. All models with TwinDISPLAY feature the intuitive control and logging software, AtmoCONTROL, which uses drag and drop symbols to input values. The software allows online monitoring of up to 32 connected appliances and can send an automatic alarm message to one or several e-mail addresses. Complex ramping processes can be programmed quickly and easy for TwinDISPLAY models. The SetpointWAIT function ensures the process time does not start before the set temperature is reached. This feature can also be used with additional, freely-positionable PT100 sensors, so that the process time starts when the set temperature is reached at all measuring points.
In aesthetic textured stainless steel and available in 3 model sizes, the ICH series is suitable for any laboratory. Unit includes: stainless steel perforated shelf, TwinDISPLAY touchscreen controller, 2.5 L water tank and connections, AtmoCONTROL Software (USB) and manual, ICH user manual, calibration certificate, power cord, and a 3-year limited manufacturers warranty.
Certifications: Standard units are safety-approved and bear the test marks: CE and EAC
Delivery information: Stainless steel grid shelving, 40 mm entry port (rear wall), Calibration Certificate, and water tank with connection for humidification system are included as standard equipment. Additional accessories or factory fitted options are not included and may affect lead time. Please contact Avantor for further details prior to ordering as these can not be retro-fitted.
Packaging: Packaged in a triple-walled corrugated box, atop a pallet.
Caution: Distilled water, used in devices with steam generator, should fulfill VDE 0510 / DIN EN 50272, and need to have a conductivity of maximal 10 μS/cm as well a pH-value between 5.0 to 7.0.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 10861-360
  • 10861-362
  • 10861-364
  • 10861-366
  • 10861-368
  • 10861-370
  • Temperature range:
  • –10 °C up to 60 °C without humiditiy; 10 °C up to 60 °C with humidity
  • –10 °C up to 60 °C without humiditiy; 10 °C up to 60 °C with humidity
  • –10 °C up to 60 °C without humiditiy; 10 °C up to 60 °C with humidity
  • –10 °C up to 60 °C without humiditiy; 10 °C up to 60 °C with humidity
  • –10 °C up to 60 °C without humiditiy; 10 °C up to 60 °C with humidity
  • –10 °C up to 60 °C without humiditiy; 10 °C up to 60 °C with humidity
  • Environmentally Preferable:
  • Sustainable Packaging|Low Manufacturing Impact
  • Sustainable Packaging|Low Manufacturing Impact
  • Sustainable Packaging|Low Manufacturing Impact
  • Sustainable Packaging|Low Manufacturing Impact
  • Sustainable Packaging|Low Manufacturing Impact
  • Sustainable Packaging|Low Manufacturing Impact
  • Control range:
  • Temperature range: Min 5 °C above ambient up to 50 °C; Humidity range: 40 to 97 % rh (and rh Off option)
  • Temperature range: Min 5 °C above ambient up to 50 °C; Humidity range: 40 to 97 % rh (and rh Off option)
  • Temperature range: Min 5 °C above ambient up to 50 °C; Humidity range: 40 to 97 % rh (and rh Off option)
  • Temperature range: Min 5 °C above ambient up to 50 °C; Humidity range: 40 to 97 % rh (and rh Off option)
  • Temperature range: Min 5 °C above ambient up to 50 °C; Humidity range: 40 to 97 % rh (and rh Off option)
  • Temperature range: Min 5 °C above ambient up to 50 °C; Humidity range: 40 to 97 % rh (and rh Off option)
  • Environmental certification:
  • EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
  • EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
  • EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
  • EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
  • EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
  • EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Volume
    Exterior Dimensions
    Electrical
    Availability
    Price
  • Climate Chamber ICH
    108 L (3.8 cu. ft)
    29.3L×25W×48.5" H
    115 V, 50/60 Hz
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (108l)
    Each (108l) - $27,596.66
    $27,596.66
    Each (108l)
       $27,596.66
    Minimum order quantity is 1
    Supplier Number: ICH110115V
    Spécifications:
    Minority or Woman-Owned Business Enterprise
      Sustainable Packaging|Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    10861-360
    Temperature range
    –10 °C up to 60 °C without humiditiy; 10 °C up to 60 °C with humidity
    Environmentally Preferable
    Sustainable Packaging|Low Manufacturing Impact
    Control range
    Temperature range: Min 5 °C above ambient up to 50 °C; Humidity range: 40 to 97 % rh (and rh Off option)
    Environmental certification
    EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
    Minority or Woman-Owned Business Enterprise
      Sustainable Packaging|Low Manufacturing Impact
  • Climate Chamber ICH
    108 L (3.8 cu. ft)
    29.3L×25W×48.5" H
    230 V, 50/60 Hz
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (108l)
    Each (108l) - $27,744.46
    $27,744.46
    Each (108l)
       $27,744.46
    Minimum order quantity is 1
    Supplier Number: ICH110230V
    Spécifications:
    Minority or Woman-Owned Business Enterprise
      Sustainable Packaging|Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    10861-362
    Temperature range
    –10 °C up to 60 °C without humiditiy; 10 °C up to 60 °C with humidity
    Environmentally Preferable
    Sustainable Packaging|Low Manufacturing Impact
    Control range
    Temperature range: Min 5 °C above ambient up to 50 °C; Humidity range: 40 to 97 % rh (and rh Off option)
    Environmental certification
    EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
    Minority or Woman-Owned Business Enterprise
      Sustainable Packaging|Low Manufacturing Impact
  • Climate Chamber ICH
    256 L (9 cu. ft)
    32.4L×28.9W×61.1" H
    115 V, 50/60 Hz
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (256l)
    Each (256l) - $39,612.42
    $39,612.42
    Each (256l)
       $39,612.42
    Minimum order quantity is 1
    Supplier Number: ICH260115V
    Spécifications:
    Minority or Woman-Owned Business Enterprise
      Sustainable Packaging|Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    10861-364
    Temperature range
    –10 °C up to 60 °C without humiditiy; 10 °C up to 60 °C with humidity
    Environmentally Preferable
    Sustainable Packaging|Low Manufacturing Impact
    Control range
    Temperature range: Min 5 °C above ambient up to 50 °C; Humidity range: 40 to 97 % rh (and rh Off option)
    Environmental certification
    EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
    Minority or Woman-Owned Business Enterprise
      Sustainable Packaging|Low Manufacturing Impact
  • Climate Chamber ICH
    256 L (9 cu. ft)
    32.4L×28.9W×61.1" H
    230 V, 50/60 Hz
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (256l)
    Each (256l) - $39,612.42
    $39,612.42
    Each (256l)
       $39,612.42
    Minimum order quantity is 1
    Supplier Number: ICH260230V
    Spécifications:
    Minority or Woman-Owned Business Enterprise
      Sustainable Packaging|Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    10861-366
    Temperature range
    –10 °C up to 60 °C without humiditiy; 10 °C up to 60 °C with humidity
    Environmentally Preferable
    Sustainable Packaging|Low Manufacturing Impact
    Control range
    Temperature range: Min 5 °C above ambient up to 50 °C; Humidity range: 40 to 97 % rh (and rh Off option)
    Environmental certification
    EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
    Minority or Woman-Owned Business Enterprise
      Sustainable Packaging|Low Manufacturing Impact
  • Climate Chamber ICH
    749 L (26.4 cu. ft)
    48.2L×32.8W×76.8" H
    115 V, 50/60 Hz
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (749l)
    Each (749l) - $53,872.38
    $53,872.38
    Each (749l)
       $53,872.38
    Minimum order quantity is 1
    Supplier Number: ICH750115V
    Spécifications:
    Minority or Woman-Owned Business Enterprise
      Sustainable Packaging|Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    10861-368
    Temperature range
    –10 °C up to 60 °C without humiditiy; 10 °C up to 60 °C with humidity
    Environmentally Preferable
    Sustainable Packaging|Low Manufacturing Impact
    Control range
    Temperature range: Min 5 °C above ambient up to 50 °C; Humidity range: 40 to 97 % rh (and rh Off option)
    Environmental certification
    EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
    Minority or Woman-Owned Business Enterprise
      Sustainable Packaging|Low Manufacturing Impact
  • Climate Chamber ICH
    749 L (26.4 cu. ft)
    48.2L×32.8W×76.8" H
    230 V, 50/60 Hz
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (749l)
    Each (749l) - $53,872.38
    $53,872.38
    Each (749l)
       $53,872.38
    Minimum order quantity is 1
    Supplier Number: ICH750230V
    Spécifications:
    Minority or Woman-Owned Business Enterprise
      Sustainable Packaging|Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    10861-370
    Temperature range
    –10 °C up to 60 °C without humiditiy; 10 °C up to 60 °C with humidity
    Environmentally Preferable
    Sustainable Packaging|Low Manufacturing Impact
    Control range
    Temperature range: Min 5 °C above ambient up to 50 °C; Humidity range: 40 to 97 % rh (and rh Off option)
    Environmental certification
    EU Directives 2002/95/EC (RoHS) and 2002/96/EC (WEEE)
    Minority or Woman-Owned Business Enterprise
      Sustainable Packaging|Low Manufacturing Impact