Compressor Cooled Incubators, ICP, Memmert
 :   Memmert
Compressor Cooled Incubators, ICP, Memmert
 :  Memmert
Compressor Cooled Incubators,  ICP, Memmert
Compressor Cooled Incubators, ICP, Memmert
Compressor-Cooled Incubator ICP* with Twin DISPLAY
COMPRESSOR-COOLED INCUBATOR ICP Ideal at temperatures around zero and below! If rapid and precise alternation between heating up and cooling down times in ramp operation is required, cooled incubators with compressor cooling prove to be in peak form - yet still work extraordinarily quiet. Due to the finely adjusted control technology, temperatures exactly reach the set point values without energy-intensive bursts of power.
AtmoCONTROL Software.
 : CE Marked.
  • Catalog No:
  • 10861-326
  • 10861-416
  • 10861-418
  • 10861-420
  • 10861-422
  • 10861-424
  • 10861-426
  • 10861-428
  • Temperature range:
  • –5…60 °C
  • –5…60 °C
  • –5…60 °C
  • –5…60 °C
  • –5…60 °C
  • –5…60 °C
  • –5…60 °C
  • –5…60 °C
Product Family Options
  • Electrical
    Volume
  • 230 V
    26.5 cu. ft.
    Each (749l)
    Each (749l) - $42,341.77
    $42,341.77
    Each (749l)
       $42,341.77
     : ICP750230V
    Minority or Woman-Owned Business Enterprise
    Specifications
    Cat. No.
    10861-326
    Temperature range
    –5…60 °C
    Minority or Woman-Owned Business Enterprise
  • 115 V
    3.8 cu. ft.
    Each (108l)
    Each (108l) - $20,455.40
    $20,455.40
    Each (108l)
       $20,455.40
     : ICP110115V
    Minority or Woman-Owned Business Enterprise
    Specifications
    Cat. No.
    10861-416
    Temperature range
    –5…60 °C
    Minority or Woman-Owned Business Enterprise
  • 230 V
    3.8 cu. ft.
    Each (108l)
    Each (108l) - $20,455.40
    $20,455.40
    Each (108l)
       $20,455.40
     : ICP110230V
    Minority or Woman-Owned Business Enterprise
    Specifications
    Cat. No.
    10861-418
    Temperature range
    –5…60 °C
    Minority or Woman-Owned Business Enterprise
  • 115 V
    9.0 cu. ft.
    Each (256l)
    Each (256l) - $26,232.26
    $26,232.26
    Each (256l)
       $26,232.26
     : ICP260115V
    Minority or Woman-Owned Business Enterprise
    Specifications
    Cat. No.
    10861-420
    Temperature range
    –5…60 °C
    Minority or Woman-Owned Business Enterprise
  • 230 V
    9.0 cu. ft.
    Each (256l)
    Each (256l) - $26,232.26
    $26,232.26
    Each (256l)
       $26,232.26
     : ICP260230V
    Minority or Woman-Owned Business Enterprise
    Specifications
    Cat. No.
    10861-422
    Temperature range
    –5…60 °C
    Minority or Woman-Owned Business Enterprise
  • 115 V
    15.9 cu. ft.
    Each (449l)
    Each (449l) - $33,990.56
    $33,990.56
    Each (449l)
       $33,990.56
     : ICP450115V
    Minority or Woman-Owned Business Enterprise
    Specifications
    Cat. No.
    10861-424
    Temperature range
    –5…60 °C
    Minority or Woman-Owned Business Enterprise
  • 230 V
    15.9 cu. ft.
    Each (449l)
    Each (449l) - $33,990.56
    $33,990.56
    Each (449l)
       $33,990.56
     : ICP450230V
    Minority or Woman-Owned Business Enterprise
    Specifications
    Cat. No.
    10861-426
    Temperature range
    –5…60 °C
    Minority or Woman-Owned Business Enterprise
  • 115 V
    26.5 cu. ft.
    Each (749l)
    Each (749l) - $42,341.77
    $42,341.77
    Each (749l)
       $42,341.77
     : ICP750115V
    Minority or Woman-Owned Business Enterprise
    Specifications
    Cat. No.
    10861-428
    Temperature range
    –5…60 °C
    Minority or Woman-Owned Business Enterprise