Histology Control Slides, Acid Fast Bacteria, Springside Scientific
100% Human Tissue.
Two stained controls in every box. Top-quality, pure white glass slides/coverglass.
: Packaged 22 (20 controls plus 2 stained) or 100 (98 controls plus 2 stained) slides in a 25-postion or 100-position plastic slide-box.
Frequently Bought Together