To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Campy, Blood Free, Karmali Agar Plate
- Size:15 x 100 mm
- Cat. no.:75832-740
- Supplier No.:G06
Campy, Blood Free, Karmali Agar for Campylobacter contains charcoal and CVA antibiotics.
Karmali formula, with charcoal, deoxycholate, cefoperazone, vancomycin, and amphotericin. Plates are designed for the selective isolation of Campylobacter spp. The Karmali formula has been shown to achieve superior results in the recovery of Campylobacter from stool specimens. Also known as mCCDA (Modified Charcoal Cefoperazone Deoxycholate Agar and Campy CSM).