ContactUsLinkComponent
HardyDisks™ AST X Factor HardyDisk™, Hemin Disk, Hardy Diagnostics
Catalog # 75832-738
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
HardyDisks™ AST X Factor HardyDisk™, Hemin Disk, Hardy Diagnostics
Catalog # 75832-738
Supplier:  Hardy Diagnostics
Supplier Number:  Z7031
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Pack type:
    1×50 disks
  • Description:
    X Factor HardyDisk™, Hemin Disk
  • Cat. no.:
    75832-738
  • Supplier No.:
    Z7031
Specifications
About this item
X Factor HardyDisk™, Hemin disk is designed to aid in the identification of Haemophilus spp.
Contains Hemin (X factor) for the speciation of Haemophilus. Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispenser by BD BioSciences. The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP), and have achieved ISO 13485 certification as a medical device manufacturer.