ContactUsLinkComponent
Cut Down Tray, Sklar
  75817-524
 :  Sklar
undefined
Cut Down Tray, Sklar
  75817-524
 :  Sklar
 :  96-1747
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Cut Down Tray
  • Cat. no.:
    75817-524
  • Packaging:
    10/Case
Cut Down Trays are industry standard
Each tray includes the following instruments and prep materials: #11 Long Scalpel, #15 Long Scalpel, OR Scissors, Straight Hemostat, Dressing Forceps, (2) Straight Mosquito Forceps, Hemostats, (3) Curved Mosquito Hemostats, Adson Tissue Forceps, Mayo-Hegar Needle Holder, Webster Needle Holder, (2) Blue Linen OR Towels, (15) Gauze Sponges (4x4", 12-ply), (2) Instrument Trays, and Clear Utility Drape.