ContactUsLinkComponent
Minor Prep Tray, Sklar
Catalogue # 75817-514
Fournisseur:  Sklar
undefined
Minor Prep Tray, Sklar
Catalogue # 75817-514
Fournisseur:  Sklar
Supplier Number:  96-1724
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Minor Prep Tray
  • Cat. no.:
    75817-514
Spécifications
À propos de cet article
Minor Prep Trays are industry standard
Each tray includes the following prep materials: PVP Prep Pad, Alcohol Prep Pad, (4) Gauze Sponges (4x4", 12-ply), and 16oz. Bowl.