Tamperproof Specimen Containers, Simport Scientific
Fournisseur: Simport Scientific
Certificats
À propos de cet article
Container and closure are made of polypropylene.
- Tight-fitting lids prevent leaks and odors
- Resist temperatures up to 121 °C
These disposable containers are ideal for collection, transport and storage of fluids, powders, solids, pathology specimens and hazardous samples. They incorporate a tamperproof lid which is opened by first removing tab on side of lid, then by lifting the cover.
Frequently Bought Together