Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Molarity Lab Investigation
- No. of Lab Groups:15
- Cat. no.:470220-224
- Supplier No.:IS2519
Designed to Introduce Students to the Concept of Molarity.
- Practice proper lab techniques
- Visually determine the unknowns
Students will first try to make a solution with a specific molarity demonstrating the importance of good laboratory technique. In the second set of experiments students will perform a titration on a known acid solution before using the concepts they have learned to identify the concentrations in three unknown solutions.