SP Wilmad-LabGlass Round Bottom Four-Neck Flasks, SP Industries
Available in various sizes, these borosilicate glass round bottom flasks have four vertical necks - one center neck and three side necks which are 90° apart
All necks have Standard Taper outer joints.
Frequently Bought Together