Sterile Polynit Heatseal LE Wipes, Contec®
Supplier: Contec
Certificates
About this item
Polynit heatseal low endotoxin wipes are manufactured from high quality 100% knitted polyester fabric with sealed edges and produce low levels of particles and fibers. The polyester fabric offers the widest range of solvent compatibility. The wipes have a guaranteed endotoxin level of <1 EU/wipe and are validated sterile to 10⁻⁶ Sterility Assurance Level, making them ideal for use in product contact areas.
- Recommended for Grade A/B or ISO Class 5 environments - product contact areas
- Laser sealed edges reduces fiber generation from the edge of the wipe
- Smaller number of wipes per bag eliminates waste
The wipes are half-folded and triple packaged in linear tear outer bags for ease of transfer into ISO Class 5 (Grades A/B) cleanrooms. This smaller packaging takes up less space, making it ideal for any size cleanroom. The small quantity of wipes per package can be used during one cleaning session, eliminating waste.
Frequently Bought Together