National Elementary Compound Microscope
Monocular Compound Microscope
Catalog # 470222-462
Supplier: National Optical
Fournisseur #: 102
Jump to:
Product Details & Documents
This low-cost elementary microscope is ideal for beginners.
- Dual LED Lighting
- Rechargeable
- Three Rechargeable NiMH Batteries Included
- One-Year Manufacturer's Warranty
- Large Focusing Knob
The cordless feature permits in and out of classroom use.
Dual LED illumination allows users to view micro specimens or larger macro objects at a lower power while the large focusing knob provides ease of use.
Ordering information: This product includes a manufacturer's warranty.
Spécifications
- Désignation:Monocular Compound Microscope
- Cat. No.:470222-462
- Illumination:LED Cordless
- Magnification:4X, 10X, 40X Objectives