Boreal Research Microscope Series
Boreal Research Microscope Series
Supplier:   VWR
Boreal Research Microscope Series
Supplier:  VWR
Certificates
Boreal Research Microscope Series
5 Options Available Below
Product Details & Documents
This microscope is ideal for advanced studies and laboratory use.
  • Choice of Objective Lenses: ASC or Semi-Plan
  • LED Illumination
  • Large, Built-In, Low-Profile, Rack-Less Stage
  • Manufacturer's Lifetime Warranty
  • Superb Optical Clarity
  • Color-Coded Objectives
The new Research series incorporates a wide range of features designed for university and laboratory use. Superb optical clarity and reliable mechanical engineering assure years of top performance.
Two grades of objective lenses are available, both of which are parfocal, parcentered, and color coded.
Ordering information: This product includes a manufacturer's warranty.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 470222-446
  • 470222-444
  • 470222-450
  • 470222-448
  • 470103-560
  • Magnification:
  • 4X, 10X, 40X, 100X Objectives
  • 4X, 10X, 40X, 100X Objectives
  • 4X, 10X, 40X, 100X Objectives
  • 4X, 10X, 40X, 100X Objectives
  • Stage:
  • Rackless Stage
  • Rackless Stage
  • Rackless Stage
  • Rackless Stage
  • Illumination:
  • LED Corded
  • LED Corded
  • LED Corded
  • LED Corded
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Binocular Compound Microscope, ASC
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,683.60
    $1,683.60
    Each
       $1,683.60
    Supplier Number: 168-ASC
    Spécifications:
    More
    Specifications
    Cat. No.
    470222-446
    Magnification
    4X, 10X, 40X, 100X Objectives
    Stage
    Rackless Stage
    Illumination
    LED Corded
  • Binocular Compound Microscope, Semi-Plan
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,580.20
    $1,580.20
    Each
       $1,580.20
    Supplier Number: 168-SP
    Spécifications:
    More
    Specifications
    Cat. No.
    470222-444
    Magnification
    4X, 10X, 40X, 100X Objectives
    Stage
    Rackless Stage
    Illumination
    LED Corded
  • Trinocular Compound Microscope, ASC
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,148.00
    $2,148.00
    Each
       $2,148.00
    Supplier Number: 169-ASC
    Spécifications:
    More
    Specifications
    Cat. No.
    470222-450
    Magnification
    4X, 10X, 40X, 100X Objectives
    Stage
    Rackless Stage
    Illumination
    LED Corded
  • Trinocular Compound Microscope, Semi-Plan
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,044.60
    $2,044.60
    Each
       $2,044.60
    Supplier Number: 169-SP
    Spécifications:
    More
    Specifications
    Cat. No.
    470222-448
    Magnification
    4X, 10X, 40X, 100X Objectives
    Stage
    Rackless Stage
    Illumination
    LED Corded
  • Micrometer Eyepiece
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $45.20
    $45.20
    Liquidation
    Each
       $45.20
    Supplier Number: SG02S0126B
    Spécifications:
    More
    Specifications
    Cat. No.
    470103-560
    Magnification
    Stage
    Illumination