Difco™ Tryptic Soy Agar, BD Biosciences
Supplier: BD
Certificates
About this item
Tryptic Soy Agar is used for the isolation and cultivation of nonfastidious and fastidious microorganisms.
- Casein and soy peptones promote general growth
- Sodium chloride maintains the osmotic equilibrium
Tryptic Soy Agar is not the medium of choice for anaerobes.
Frequently Bought Together