Difco™ Tetrathionate (TT) Broth Base, Hajna, BD Biosciences
Catalog # CA90003-424
Supplier: BD
CAS Number:
Specifications
- Description:Difco™ Tetrathionate (TT) Broth Base, Hajna
- Size:500 g
- Cat. no.:CA90003-424
- Supplier No.:249120
Specifications
About this item
TT Broth Base, Hajna is used for enriching Salmonella from food and dairy products prior to isolation procedures.
- Peptone provides nitrogen and amino acids
- Yeast extract supplies growth factors and vitamins
- Dextrose and mannitol are fermentable carbohydrates
- Calcium carbonate is a neutralizer that absorbs toxic metabolites
Organisms containing the enzyme tetrathionate reductase will proliferate in this medium.