Tryptone Azolectin Tween (TAT) Broth Base, BD Biosciences
Catalogue # CA90005-136
Fournisseur: BD
Numéro CAS:
Spécifications
- Description:Tryptone Azolectin Tween (TAT) Broth Base
- Size:500 g
- Cat. no.:CA90005-136
- Supplier No.:298410
Spécifications
À propos de cet article
TAT Broth Base is used for cultivating microorganisms from highly viscous or gelatinous materials.
- Peptone provides nitrogen, vitamins, amino acids, and carbon
- Especially adapted to the testing of cosmetics
Soy Lecithin and polysorbate 20 neutralize preservatives in the cosmetics or pharmaceutical products, allowing bacteria to grow.