Difco™ Supplement VX with Reconstituting Fluid, BD Biosciences
Fournisseur: BD
Certificats
À propos de cet article
Supplement VX with Reconstituting Fluid is used for supplementing media to culture fastidious microorganisms, particularly Neisseria gonorrhoeae and Haemophilus influenzae.
- Lyophilized concentrate
- Contains vitamins, amino acids, coenzymes, and dextrose
This supplement is recommended for enriching GC Agar media, Proteose No. 3 Agar, and the selective agars for the isolation of pathogenic Neisseria.
Frequently Bought Together