Difco™ Modified mTEC Agar, BD
Fournisseur: BD
Certificats
À propos de cet article
Modified mTEC Agar is a selective culture medium used for the chromogenic detection and enumeration of thermotolerant Escherichia coli in water by the membrane filtration technique.
- Peptone is a source of nitrogen, amino acids, and carbon
- Agar is the solidifying agent
Sodium lauryl sulfate and sodium desoxycholate are selective against Gram-positive bacteria. Red to magenta colonies may be presumptively identified as
E. coli.
Frequently Bought Together