Difco™ Select APS™ Super Broth Base, BD Diagnostics | Avantor
Difco™ Select APS™ Super Broth Base, BD Diagnostics
Supplier:   BD
Difco™ Select APS™ Super Broth Base, BD Diagnostics
Supplier:  BD
Certificates
Difco™ Select APS™ Super Broth Base, BD Diagnostics
2 Options Available Below
Product Details & Documents
Select APS™ Super Broth is a molecular genetics medium used to grow Escherichia coli to a high cell density.
  • Molecular genetics medium
  • Alternative to classical animal-based media
  • Allows maintenance and propogation of E. coli
  • Soy hydrolysate provides nitrogen and carbon
  • Glycerol is added as a carbon and energy source
Select APS™ media are manufactured from animal-free ingredients in order to minimize the risk of bovine spongiform encephalopathy in culture media containing animal, and especially bovine, materials.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Difco™ Select APS™ Super Broth Base
    10 kg
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (10kg)
    Each (10kg) - $2,810.00
    $2,810.00
    Each (10kg)
       $2,810.00
    Supplier Number: 212486
  • Difco™ Select APS™ Super Broth Base
    500 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (500g)
    Each (500g) - $240.00
    $240.00
    Each (500g)
       $240.00
    Supplier Number: 212485