m Staphylococcus Broth, BD Life Sciences
m Staphylococcus Broth, 500 g
  CA90003-766
 :   BD
 :  264920
m Staphylococcus Broth, BD Life Sciences
  CA90003-766
 :  BD
 :  
m Staphylococcus Broth, BD Life Sciences
m Staphylococcus Broth, 500 g
 :  
$414.00
Each (500g)
m Staphylococcus Broth is used for isolating staphylococci by the membrane filtration technique.
  • Staphylococci are indicators of recreational water quality
  • Used in a multiple-tube procedures
  • For monitoring swimming pool water
  • For the identification of S. aureus
  • Dehydrated appearance: Light beige, free-flowing
Observe membranes for growth and pigment production. Test for mannitol fermentation by adding a drop of bromthymol blue to the site from which a colony is removed; a yellow color indicates mannitol fermentation.
  • Désignation:
    m Staphylococcus Broth
  • Taille:
    500 g