PHCbi's VIP® Series ultra-low temperature chest freezers are designed for safe, reliable storage of the most critical biologicals.
- Reliable
- Maximum storage
- Minimal footprint
- VIP Plus insulation composite
- Uniform preservation
- Microprocessor controls and alarms
- Five-year manufacturer's parts and labor warranty
Energy efficiency in VIP® Series freezers is achieved through balanced temperature performance and power management
The robust refrigeration system is powered by PHCbi-engineered compressors. The system provides dependable cooling by balancing temperature performance and energy management. Reserve refrigeration power assures exceptional temperature recovery following door openings. VIP® freezers have demonstrated >99% reliability with excellent performance history.
The advanced combination of cabinet design, electronics, refrigeration, and critical components are engineered for enhanced security, performance, and reliability.
- Catalog No:
- CA75788-512
- Model:
- MDFDC700VXCPA
- Cat. No.:
- 75788-512
- Insulation:
- Rigid Polyurethane Foamed-In Place
- Electrical:
- 208/230 V
- Weight:
- 358 kg (848.8 lbs.)