Biox Cell Spreaders
Biox Cell Spreaders
Biox Cell Spreaders
Biox Cell Spreaders
Cell Spreaders are ideal for the even spreading of bacterial cultures.
  • Polystyrene construction
  • L-Shape design
  • Disposable
  • Smooth spreading surface with slight upward angle reduces the risk of damaging agar surfaces
  • Gamma radiation sterilized
  • Individually wrapped
L-Shaped cell spreaders allows for application on the entire culture dish or plate surface.
Product Family Options
  • Color
    Packaging
  • Orange
    Individually Wrapped
    Case of 500
    Case of 500 - $226.43
    $226.43
    Case of 500
       $226.43
     : BX65-1001
  • Orange
    Bulk (10/pack)
    Case of 500
    Case of 500 - $203.72
    $203.72
    Case of 500
       $203.72
     : BX65-1010