Biox Cell Strainers
Biox Cell Strainers
Biox Cell Strainers
Biox Cell Strainers
Cell Strainers feature a strong nylon mesh for optimal performance in a variety of applications.
  • Polypropyelene frame with nylon mesh
  • Gamma radiation sterilized
  • Color coded for easy pore size identification
  • Individually wrapped
  • Applications in cell culture and filtration of various cells
Perfect fit for 50 ml centrifuge tubes and color coded according to the pore size.
Product Family Options
  • Taille de pore
    Color
  • 40 µm
    Purple
    Case of 100
    Case of 100 - $331.70
    $331.70
    Case of 100
       $331.70
     : BX15-1040
  • 70 µm
    Orange
    Case of 100
    Case of 100 - $404.60
    $404.60
    Case of 100
       $404.60
     : BX15-1070
  • 100 µm
    Green
    Case of 100
    Case of 100 - $393.59
    $393.59
    Case of 100
       $393.59
     : BX15-1100