Biox Cell Scrapers and Lifters
Supplier: TWD Scientific
Certificates
About this item
Cell Scrapers feature a uniquely designed handle and blade which reduces cell damage during the harvesting process.
- Non-pyrogenic
- Polyethylene construction
- Pivoting blade makes precise cutting and scraping movements
- Disposable
- Pre-sterilized and individually wrapped
Our scrapers have pivot blades for precise cutting and scraping and our lifters are one sturdy piece for improved control.
Frequently Bought Together