Biox Freezer Boxes
Supplier:   TWD Scientific
Biox Freezer Boxes
Supplier:  TWD Scientific
Certificates
Biox Freezer Boxes
Biox Freezer Boxes
4 Options Available Below
Product Details & Documents
Freezer Boxes are designed to provide easy inventory management of cryogenic vials used in tissue culture, pharmaceutical research, and other biotechnology labs.
  • Store in mechanical freezers (PC/PP) or liquid nitrogen (PC)
  • Vented boxes allow rapid cooling and full drainage
  • Chemically resistant to alcohols and mild solvents
  • Grid on lid for easy identification
Plastic freezer boxes are specifically designed for Liquid nitrogen (PC) and mechanical freezer (PP) storage.
Product Family Options
Informations sur le produit
  • Désignation
    Nb de places
    Hauteur
    Availability
    Price
  • Freezer Box, Polycarbonate
    81
    5 cm (2")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $387.80
    $387.80
    Case of 20
       $387.80
    Supplier Number: BX90-9009-1
  • Freezer Box, Polycarbonate
    100
    5 cm (2")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $382.27
    $382.27
    Case of 20
       $382.27
    Supplier Number: BX90-9210-1
  • Freezer Box, Polypropylene
    81
    5 cm (2")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $170.35
    $170.35
    Case of 20
       $170.35
    Supplier Number: BX90-9081
  • Freezer Box, Polypropylene
    100
    5 cm (2")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 20
    Case of 20 - $238.59
    $238.59
    Case of 20
       $238.59
    Supplier Number: BX90-9100