ContactUsLinkComponent
Biox Tri-Cornered Beakers
  75799-910
 : BX93-0050
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Biox Tri-Cornered Beakers
  75799-910
 :  BX93-0050
 : BX93-0050
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Select an Option
50 ml (1.7 oz.)
Select an Option
Clear
Select an Option
75799-910
Select an Option
93-0050
Tri-Cornered Beakers are designed to provide three dripless pouring spouts
  • Strong medical-grade polypropylene construction
  • Equipped with three dripless pouring spouts
  • Good impact strength
  • Translucent and autoclavable
  • Disposable, easy to clean, and durable enough for reuse
  • Molded Graduations
Available in six different sizes.
Frequently Bought Together