TWD Barcoded Autosampler Vials
TWD Barcoded Autosampler Vials
TWD Barcoded Autosampler Vials
TWD Barcoded Autosampler Vials
TWD Barcoded Autosampler Vials with 2D barcode.
  • Vials feature writing patch and graduations
  • Glass construction
  • Screw-top 9-425 GPI thread finish
TWD barcoded autosampler kits are a cost-effective option if you're looking for a easy, hassle-free option to track your samples your samples.
 : Convenient Kit includes black PP cap and red rubber/PTFE Septa; available as vial and cap kit or cap only.
Product Family Options
  • Désignation
    Capacité
    Diameter ×H
    Matière
    Color
    Avec bouchon
    Septum
  • Vial without cap
    2.00 ml
    12×32 mm
    Glass
    Amber
    Non
    Case of 100
    Case of 100 - $121.23
    $121.23
    Case of 100
       $121.23
     : GV1232A-P-2DLE
  • Vial without cap
    2.00 ml
    12×32 mm
    Glass
    Clear
    Non
    Case of 100
    Case of 100 - $114.32
    $114.32
    Case of 100
       $114.32
     : GV1232C-P-2DLE
  • Vial and cap kit
    2.00 ml
    12×32 mm
    Glass
    Amber
    Oui
    Red rubber/PTFE
    Case of 100
    Case of 100 - $196.06
    $196.06
    Case of 100
       $196.06
     : GV1232A-P-2DK
  • Vial and cap kit
    2.00 ml
    12×32 mm
    Glass
    Clear
    Oui
    Red rubber/PTFE
    Case of 100
    Case of 100 - $192.70
    $192.70
    Case of 100
       $192.70
     : GV1232C-P-2DK
  • Cap only, black
     : -