BBL™ Vitamin K1-Hemin Solution, BD Biosciences
Catalog # CA90000-618
Supplier: BD
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:BBL™ Vitamin K1-Hemin Solution
- Size:10 mL
- Environmentally Preferable:
- Cat. no.:CA90000-618
- Supplier No.:212354
Specifications
About this item
Vitamin K1-Hemin Solution is used as a culture medium enrichment for anaerobic microorganisms.
- Ready-to-use solution
- Must be added aseptically
- Employed in prepared media
Vitamin K1-Hemin Solution is used in anaerobic media.