Order Entry
Canada
ContactUsLinkComponent
BBL™ Mycoplasma Broth Base (Frey), BD Biosciences
BBL™ Mycoplasma Broth Base (Frey), BD Biosciences
Catalog # CA90000-610
Supplier:  BD
CAS Number:  
BBL™ Mycoplasma Broth Base (Frey), BD Biosciences
Catalog # CA90000-610
Supplier:  BD
Supplier Number:  212346
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    BBL™ Mycoplasma Broth Base (Frey)
  • Size:
    500 g
  • Environmentally Preferable:
  • Cat. no.:
    CA90000-610
  • Supplier No.:
    212346

Specifications

About this item

Mycoplasma Broth Base, when supplemented with nutritive enrichments, are used for isolating and cultivating Mycoplasma.

  • Supplemented by mycoplasma enrichment without penicillin
  • Contains fresh yeast extract and horse serum
  • Specially developed for the cultivation of avian strains of Mycoplasma

Mycoplasma Broth Base (Frey) is used for the cultivation of avian mycoplasmas.