Boreal Compound Microscopes with 8" LCD Tablet | Avantor
Boreal Compound Microscopes with 8" LCD Tablet
Supplier:   VWR International
Boreal Compound Microscopes with 8" LCD Tablet
Supplier:  VWR International
Certificates
Boreal Compound Microscopes with 8" LCD Tablet
Boreal Compound Microscopes with 8" LCD Tablet
2 Options Available Below
Product Details & Documents
Use it as a traditional microscope, or view and share live images with the tablet screen.
  • LCD Android Tablet, Eight Inches
  • Screen Resolution: 800×1280
  • Built-in Motic Hub for Wireless Sharing
  • Motic Images Plus Software Included
These microscopes combine high speed imaging technology with our most popular microscopes.
The EM model is coarse focus only and includes 4X, 10X and 40X objectives and gliding stage with stage clips. The HM model has coarse and fine focus, 4X, 10X, 40X and 100X objectives and a built-in mechanical stage. Both models have LED corded illumination.
The Wi-Fi digital tablets transmit live images to iOS or Android devices. Use them as conventional microscopes or share live images with colleagues using Wi-Fi tablets or wireless laptops. The tablet includes preloaded Motic apps.
Ordering information: This kit includes a lifetime manufacturer's warranty on the microscope chassis and a one-year warranty on the tablet.
Product Family Options
Informations sur le produit
  • Désignation
    Grossissement
    Illumination
    Availability
    Price
  • HM series microscope
    4X, 10X, 40X, 100X Objectives
    LED corded
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 2 055,35 $
    2 055,35 $
    Each
       2 055,35 $
  • EM series microscope
    4X, 10X, 40X Objectives
    LED corded
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 1 551,35 $
    1 551,35 $
    Each
       1 551,35 $