Repeater® E3 and E3x Pipettors, Eppendorf® | Avantor
Repeater® E3 and E3x Pipettors, Eppendorf®
Repeater® E3 and E3x Pipettors, Eppendorf®
Supplier:  Eppendorf
Certificates
Repeater® E3 and E3x Pipettors, Eppendorf®
Repeater® E3 and E3x Pipettors, Eppendorf®
2 Options Available Below
Product Details & Documents
The Repeater® E3/E3x Pipettors, in combination with the Combitips Advanced® pipette tips, creates a versatile dispensing system that functions according to the positive displacement principle.
  • Automatic Combitip® recognition
  • Ergonomic one-button tip ejector
  • Precisely programmable aspiration and dispensing speeds
  • Repetitive dispensing modes
  • Wide dispensing range: 1 μl to 50 ml, in 100-nanoliter increments
  • Safe and ergonomic operation with 9 different selectable languages
These devices provide precise and accurate dispensing of viscous solutions (e.g. glycerol) or liquids with high vapor pressure (e.g. ethanol). Contamination-free dispensing for toxic, radioactive, and other hazardous liquids (all available modes). Large illuminated color display reduces eye strain when reading. Motor-driven function reduces human error to a minimum, and reduces the risk of repetitive strain injuries (RSI).
It is therefore capable of pipetting any liquid while maintaining the highest standards for accurate results.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Repeater® E3 with Power Cable, Combitips® Pack
    Discontinued
    Supplier Number: 4987000118
  • Repeater® E3 Bundle, Including Charger Stand
    Discontinued
    Supplier Number: 4987000398
  • Repeater® E3x with Power Cable, Combitips® Pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,970.00
    $2,970.00
    Each
       $2,970.00
    La quantité minimale de commande est de 1
    Supplier Number: 4987000134
  • Repeater® E3x Bundle, Including Charger Stand
    Discontinued
    Supplier Number: 4987000410
  • Repeater® E3 Bundle, Including Charger Stand
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,780.00
    $2,780.00
    Each
       $2,780.00
    Supplier Number: 4987000398
  • Repeater® E3x Bundle, Including Charger Stand
    Discontinued
    Supplier Number: 4987000410