Microbank™-Dry, Pro-Lab Diagnostics | Avantor
Microbank™-Dry, Pro-Lab Diagnostics
Marketsource
Microbank™-Dry, Pro-Lab Diagnostics
Marketsource
Certificates
Microbank™-Dry, Pro-Lab Diagnostics
Microbank™-Dry, Pro-Lab Diagnostics
3 Options Available Below
Product Details & Documents
A ready-to-use system designed to simplify the storage and retrieval of bacterial and fungal cultures.
  • Longer survival of fastidious cultures
  • Higher quantitative recoveries
  • Utilizes porous glass beads and a specially formulated cryopreservative for storage at low temperatures
Each Microbank™ vial contains approximately 25 sterile coloured beads (single colour) and the cryopreservative
The long term storage of microorganisms is a significant challenge in microbiology. Microbank™ offers a platform that utilizes porous glass beads and a specially formulated cryopreservative for storage at low temperatures. This format coupled with the Microbank™ Cryoblock offers the least possibility of disturbance to the stored organism, yet, permits ready and rapid access.
Each Microbank™-Dry vial contains approximately 25 sterile coloured beads (single colour). The specially treated beads are of a porous nature allowing microorganisms to readily adhere onto the bead surface. After
the addition of cryopreservative and inoculation the Microbank™-Dry vials are kept at -70°C for extended storage. When a fresh culture is required, a single bead is easily removed from the Microbank™-Dry vial and used to directly inoculate a suitable culture medium.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Packaging
    Availability
    Price
  • Microbank, dry
    Blue
    EA
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 80 (2ml)
    Pack of 80 (2ml) - $631.02
    $631.02
    Pack of 80 (2ml)
       $631.02
    Supplier Number: PL.172-B
  • Microbank, dry
    Green
    EA
    Discontinued
    Supplier Number: PL.172-G
    Spécifications:
    Marketsource
    Marketsource
  • Microbank, dry
    Light blue
    EA
    Discontinued
    Supplier Number: PL.172-LB
    Spécifications:
    Marketsource
    Marketsource
  • Microbank, dry
    Assorted
    EA
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 80 (2ml)
    Pack of 80 (2ml) - $636.94
    $636.94
    Pack of 80 (2ml)
       $636.94
    Supplier Number: PL.172-M
  • Microbank, dry
    Red
    EA
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 80 (2ml)
    Pack of 80 (2ml) - $633.94
    $633.94
    Pack of 80 (2ml)
       $633.94
    Supplier Number: PL.172-R
  • Microbank, dry
    Yellow
    EA
    Discontinued
    Supplier Number: PL.172-Y
    Spécifications:
    Marketsource
    Marketsource