Pro-Loops Nichrome Wire Loops, Pro-Lab Diagnostics
Supplier: PRO-LAB DIAGNOSTICS INC CA
Certificates
About this item
When strict calibration is required, certified/calibrated loops from Pro-Lab Diagnostics are manufactured from the highest quality 80/20 nickel chromium wire.
- Useful for strict calibration
- Manufactured from the highest quality, 80/20 nickely chromium wire
- Presented in a protective case
When strict calibration is required, certified/calibrated loops from Pro-Lab Diagnostics are manufactured from the highest quality 80/20 nickel chromium wire and calibrated using the Evans Blue Dye Method. Each pack of five loops is presented in a protective case and accompanied by the appropriate certificate of calibration.
Frequently Bought Together