50X TAE (Tris-Acetate EDTA), pH 8.3
Supplier: GROWCELLS
Certificates
About this item
TAE buffer is used in agarose gel electrophoresis and Polyacrylamide gels both as a buffer and a component of a gel
Can be used for both genomic and large supercoiled DNA. TAE buffer has a relatively low buffering capacity therefore buffer should be replaced periodicly during long electrophoresis runs. Offered in different concentration for all common biological applications.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together